

N,N-Dimethylacetamid 99%, Reagent Grade
Catalog # D137510-2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:2,5 L
- Sekvence:5096
Specifications
Specification Test Results
- Vapor Pressure:2 mmHg ( 25 °C)
- Identity (IR):complying
- Assay (GC):Min. 98.5 %
- Explosion Limit:11.5 %, 160 °F
- Autoignition Temperature:914 °F
- Refractive Index:n20/D 1.437 (lit.)
- Vapor Density:3 (vs air)
Specification Test Results
About this item
:
Merck Index:13,03254
Boiling Pt:11,5 %(V) (99 °C)
Melting Pt:-20 °C
Melting Pt:–20 °C
Density:0,94 g/cm³ (20 °C)
Flash Pt:64 °C
EINECS:204-826-4
ADR:6.1,III
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:127-19-5
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686