

N,N-Dimethylacetamide ≥99.5% for HPLC
Catalog # 22916.K2
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:87.12 g/mol
Merck Index:14,3227
Boiling Pt:165…166 °C
Melting Pt:–20 °C
Density:0.940
Flash Pt:70 °C (158 °F)
Storage Temperature:Ambient
EINECS:204-826-4
UN:2810
ADR:6.1,III
:GHS08
:GHS07
CAS Number:127-19-5
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686