

N,N-Dimethylacetamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:127-19-5
:
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686
:87.12 g/mol
Merck Index:13,03254
Boiling Pt:166.1 °C (1013 hPa)
Melting Pt:–20 °C
Density:0.937 g/cm³
Flash Pt:70 °C
Storage Temperature:Ambient
EINECS:204-826-4
ADR:6.1,III
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4370 to 1.4390 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥99.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.01 %
- Free acid (CH3COOH):≤0.1 %
Specification Test Results
Frequently Bought Together