Zadání objednávky
Czech Republic
ContactUsLinkComponent
4,4-Dimethoxybutanone 92%
Flammable
4,4-Dimethoxybutanone 92%
Certificates
undefined
Flammable
4,4-Dimethoxybutanone 92%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4130 to 1.4160 (20°C, 589 nm)
  • GC:
    ≥90 %

Specification Test Results

Related Information

Frequently Bought Together