
Diethyl pyrocarbonate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1609-47-8
:
Formula:O(COOC₂H₅)₂
MDL Number:MFCD00009106
:162.14 g/mol
Merck Index:14,7998
Boiling Pt:93…94 °C (18 mmHg)
Density:1.101
Flash Pt:69 °C (156 °F)
EINECS:216-542-8
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together