Ethyl acetoacetate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:141-97-9
:
Formula:C₆H₁₀O₃
MDL Number:MFCD00009199
Merck Index:13,03793
Boiling Pt:180.8 °C (1013 hPa)
Melting Pt:–45 °C
Density:1.0188 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:65 °C
:130.14 g/mol
EINECS:205-516-1
:GHS07
Storage Temperature:Ambient
Frequently Bought Together