2-Tridecanone, Sigma-Aldrich®
Catalog # SIAL172839-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:198.35 g/mol
Boiling Pt:263 °C (1013 hPa)
Melting Pt:25…27 °C
Density:0.82 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:209-784-0
CAS Number:593-08-8
Formula:C₁₃H₂₆O
MDL Number:MFCD00008968