6-Chloro-1-hexanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2009-83-8
:
Formula:C₆H₁₃ClO
MDL Number:MFCD00002984
:136.62 g/mol
Boiling Pt:110 °C (14 mmHg)
Density:1.0241 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:98 °C
Storage Temperature:Ambient
EINECS:217-925-2
Frequently Bought Together