Order Entry
Czech Republic
ContactUsLinkComponent
Propylene oxide ≥99%, TECHNICAL
Flammable
Health
Irritant
Propylene oxide ≥99%, TECHNICAL
Catalog # 27165.295
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Irritant
Propylene oxide ≥99%, TECHNICAL
Catalog # 27165.295
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 99 %

Specification Test Results

About this item

 :
 :58.08 g/mol
Merck Index:12,08041
Boiling Pt:34.3 °C (1013 hPa)
Melting Pt:–112 °C
Density:0.83 g/cm³ (20 °C)
Flash Pt:–37 °C
EINECS:200-879-2
UN:1280
ADR:3,I
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Refrigerator
CAS Number:75-56-9
Formula:C₃H₆O
MDL Number:MFCD00005126