


1,2-Epoxybutane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:106-88-7
:
Formula:C₄H₈O
MDL Number:MFCD00005153
:72.11 g/mol
Boiling Pt:63 °C (1013 hPa)
Melting Pt:–130 °C
Density:0.83 g/cm³ (20 °C)
Flash Pt:–26 °C
Storage Temperature:Ambient
UN:3022
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:203-438-2
:GHS02
:GHS08
:GHS07
Specification Test Results
- Solubility:59 g/l (20 °C)
- Melting Point:-130 °C
- Molar Mass:72.1 g/mol
- Boiling Point:63 °C (1013 hPa)
- Vapor Pressure:188 hPa (20 °C)
- Flash Point:-26 °C
- Refractive Index:1.3835 (20 °C, 589 nm)
- Explosion Limit:1.7 - 19 %(V)
- Density:0.83 g/cm3 (20 °C)
- pH:7 (50 g/l, H2O, 20 °C)
- Ignition Point:370 °C
Specification Test Results
Frequently Bought Together