
3-Bromo-1-propanol 97%
:627-18-9
:
:BrH₂CCH₂CH₂OH
:MFCD00002942
:138.99 g/mol
:62 °C (5 mmHg)
:1.6 g/cm³
:65 °C
:210-986-6
:GHS07
:Refrigerator
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4860 to 1.4900 (20°C, 589 nm)
- GC:≥96.0 %
Frequently Bought Together