1,3-Dibromo-2-propanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:96-21-9
:
Formula:BrH₂CCH(OH)CH₂Br
MDL Number:MFCD00000216
:217.89 g/mol
Boiling Pt:89 °C (1013 hPa)
Density:2.11 g/cm³ (0 °C)
EINECS:202-489-8
UN:1992
ADR:3,III
:M-Clarity™ quality level = MQ100
Frequently Bought Together