
6-Bromohexanoyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22809-37-6
:
Formula:BrH₂C(CH₂)₄COOCl
MDL Number:MFCD00000761
:213.5 g/mol
Boiling Pt:119…120 °C (9 mmHg)
Density:1.397
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:245-236-7
UN:3265
ADR:8,II
:GHS05
Related Information
Frequently Bought Together