
1,5-Dibromopentane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-24-0
:
Formula:BrH₂C(CH₂)₃CH₂Br
MDL Number:MFCD00000268
:229.94 g/mol
Boiling Pt:224 °C (1013 hPa)
Melting Pt:–39 °C
Density:1.7 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:203-849-7
:GHS07
Storage Temperature:Ambient
Frequently Bought Together