Linoleic acid, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:60-33-3
:
Formula:C₁₈H₃₂O₂
MDL Number:MFCD00064241
:280.45 g/mol
Merck Index:13,05527
Boiling Pt:360 °C (1013 hPa)
Melting Pt:–5 °C
Density:0.9 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:200-470-9
Frequently Bought Together