(S)-(-)-1-Octyn-3-ol, Sigma-Aldrich®
Catalog # SIAL393967-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:126.2 g/mol
Boiling Pt:130 °C (1013 hPa)
Density:0.866 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
CAS Number:32556-71-1
Formula:C₈H₁₄O
MDL Number:MFCD00191475