


(R)-(+)-3-Butyne-2-ol 98%, 98% ee
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:42969-65-3
:
Formula:C₄H₆O
MDL Number:MFCD00211237
:70.09 g/mol
Boiling Pt:108…111 °C
Density:0.89 g/cm³
Flash Pt:6 °C
UN:2929
ADR:6.1,II
:GHS06
:GHS02
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4235 to 1.4255 (20°C, 589 nm)
- GC:≥97.5 %
- Enantiomeric excess:≥97.5 % (GC)
Specification Test Results
Frequently Bought Together