Order Entry
Czech Republic
ContactUsLinkComponent
L(+)-Valine
L(+)-Valine
Catalog # A1637.1000
Supplier:  PanReac AppliChem
CAS Number:  
L(+)-Valine
Catalog # A1637.1000
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :117.15 g/mol
Merck Index:13,09458
Boiling Pt:217 °C (1013 hPa)
Melting Pt:86 °C
Density:1.284 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-773-6
CAS Number:72-18-4
Formula:C₅H₁₁NO₂
MDL Number:MFCD00064220