(2S,3S)-(+)-2,3-Butanediol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:19132-06-0
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00063648
:90.12 g/mol
Boiling Pt:179 °C (1013 hPa)
Density:0.99 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
Frequently Bought Together