Order Entry
Czech Republic
ContactUsLinkComponent
 
(2S,3S)-(+)-2,3-Butanediol, Supelco®
Catalog # 18967-1ML
Supplier:  Merck
CAS Number:  
undefined
(2S,3S)-(+)-2,3-Butanediol, Supelco®
Catalog # 18967-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :90.12 g/mol
Boiling Pt:179 °C (1013 hPa)
Density:0.99 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
CAS Number:19132-06-0
Formula:C₄H₁₀O₂
MDL Number:MFCD00063648