(S)-(+)-2-Hexanol, Sigma-Aldrich®
Catalog # SIAL655236-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:102.18 g/mol
Boiling Pt:137 °C (1013 hPa)
Density:0.81 g/cm³ (20 °C)
ADR:3,III
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
UN:2282
CAS Number:52019-78-0
Formula:C₆H₁₄O
MDL Number:MFCD00065955