
(S)-(+)-2-Methoxy-1-propanol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:116422-39-0
:
Formula:C₄H₁₀O₂
MDL Number:MFCD01632593
:90.12 g/mol
Density:0.938 g/cm³
Flash Pt:33 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4060 to 1.4090 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥96.0 %
- Specific optical rotation:+38° to +44° (20°C, 589 nm) (c=1.66, CHCl3)
Specification Test Results
Frequently Bought Together