
L(+)-Tartaric acid 99.5-101.0% (dried basis), puriss. p.a. reag. ISO, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:87-69-4
:
Formula:C₄H₆O₆
MDL Number:MFCD00064207
Merck Index:13,09158
Boiling Pt:399 °C (1013 hPa)
Melting Pt:168…170 °C
Density:1.759 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:150 °C (closed cup)
:150.09 g/mol
Storage Temperature:Ambient
EINECS:201-766-0
REACH:01-2119537204-47
:GHS05
Frequently Bought Together