Order Entry
Czech Republic
ContactUsLinkComponent
L(+)-Tartaric acid 99.7-100.5% (dried basis), powder, puriss., meets analytical specification of Ph. Eur., NF, Sigma-Aldrich®
Corrosive
L(+)-Tartaric acid 99.7-100.5% (dried basis), powder, puriss., meets analytical specification of Ph. Eur., NF, Sigma-Aldrich®
Catalog # SIAL27506-1KG-R
Supplier:  Merck
CAS Number:  
undefined
Corrosive
L(+)-Tartaric acid 99.7-100.5% (dried basis), powder, puriss., meets analytical specification of Ph. Eur., NF, Sigma-Aldrich®
Catalog # SIAL27506-1KG-R
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
Merck Index:13,09158
Boiling Pt:399 °C (1013 hPa)
Melting Pt:168…170 °C
Density:1.759 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Flash Pt:150 °C (closed cup)
 :150.09 g/mol
Storage Temperature:Ambient
EINECS:201-766-0
REACH:01-2119537204-47
 :GHS05
CAS Number:87-69-4
Formula:C₄H₆O₆
MDL Number:MFCD00064207