Organic reference standard, 2,2',3,3',4,5,6,6'-Octachlorobiphenyl (PCB No. 199) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20019900IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:52663-73-7
- Description:2,2',3,3',4,5,6,6'-Octachlorobiphenyl (PCB No. 199) 10 µg/ml in isooctane
- Formula:C₁₂H₂Cl₈
- Description:2,2',3,3',4,5,6,6'-Octachlorobiphenyl (PCB No. 199)