Organic reference standard, 2,2',3,3',4,4',5,6,6'-Nonachlorobiphenyl (PCB No. 207) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-207
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:52663-79-3
- Description:2,2',3,3',4,4',5,6,6'-Nonachlorobiphenyl (PCB No. 207) 35 µg/ml in isooctane
- Formula:C₁₂HCl₉
- Description:2,2',3,3',4,4',5,6,6'-Nonachlorobiphenyl (PCB No. 207)