Organic reference standard, 2,2',3,3',4,4',5,5',6-Nonachlorobiphenyl (PCB No. 206) 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20020600IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:40186-72-9
- Description:2,2',3,3',4,4',5,5',6-Nonachlorobiphenyl (PCB No. 206) 10 µg/ml in isooctane
- Formula:C₁₂HCl₉
- Description:2,2',3,3',4,4',5,5',6-Nonachlorobiphenyl (PCB No. 206)