Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, 2,4,6-Trichlorobiphenyl (PCB No. 30) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20003000IO
 :  
Organic reference standard, 2,4,6-Trichlorobiphenyl (PCB No. 30) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20003000IO
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    35693-92-6
  • Description:
    2,4,6-Trichlorobiphenyl (PCB No. 30) 10 µg/ml in isooctane
  • Formula:
    C₁₂H₇Cl₃
  • Description:
    2,4,6-Trichlorobiphenyl (PCB No. 30)