Organic reference standard, 2,4,5-Trichlorobiphenyl (PCB No. 29) 35 µg/ml in isooctane, CIL
Katalogové číslo PROOCIL-PCB-29
Dodavatel: LGC Standards PROMOCHEM
Číslo CAS:
Specifikace
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:15862-07-4
- Description:2,4,5-Trichlorobiphenyl (PCB No. 29) 35 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:2,4,5-Trichlorobiphenyl (PCB No. 29)