Organic reference standard, 2,4,5-Trichlorobiphenyl (PCB No. 29) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20002900IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:15862-07-4
- Description:2,4,5-Trichlorobiphenyl (PCB No. 29) 10 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:2,4,5-Trichlorobiphenyl (PCB No. 29)