Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, 2,3,4,6-Tetrachlorobiphenyl (PCB No. 62) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,3,4,6-Tetrachlorobiphenyl (PCB No. 62) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-62
CAS Number:  
Organic reference standard, 2,3,4,6-Tetrachlorobiphenyl (PCB No. 62) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-62
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    54230-22-7
  • Description:
    2,3,4,6-Tetrachlorobiphenyl (PCB No. 62) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₆Cl₄
  • Description:
    2,3,4,6-Tetrachlorobiphenyl (PCB No. 62)

Specifications

About this item