Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, 2,3,4,5-Tetrachlorobiphenyl (PCB No. 61) 10 µg/ml in isooctane, Dr Ehrenstorfer
Organic reference standard, 2,3,4,5-Tetrachlorobiphenyl (PCB No. 61) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20006100IO
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2,3,4,5-Tetrachlorobiphenyl (PCB No. 61) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20006100IO
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    33284-53-6
  • Description:
    2,3,4,5-Tetrachlorobiphenyl (PCB No. 61) 10 µg/ml in isooctane
  • Formula:
    C₁₂H₆Cl₄
  • Description:
    2,3,4,5-Tetrachlorobiphenyl (PCB No. 61)

Specifications

About this item