Organic reference standard, 4-Chlorobiphenyl (PCB No. 3) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-3-CS
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:2051-62-9
- Description:4-Chlorobiphenyl (PCB No. 3) 100 µg/ml in isooctane
- Formula:C₁₂H₉Cl
- Description:4-Chlorobiphenyl (PCB No. 3)