Organic reference standard, 4,4'-Dichlorobiphenyl (PCB No. 15) 100 µg/ml in isooctane, CIL
PROOCIL-PCB-15-CS
:
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:2050-68-2
- Description:4,4'-Dichlorobiphenyl (PCB No. 15) 100 µg/ml in isooctane
- Formula:C₁₂H₈Cl₂
- Description:4,4'-Dichlorobiphenyl (PCB No. 15)