Organic reference standard, 3,3'-Dichlorobiphenyl (PCB No. 11) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-11
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:2050-67-1
- Description:3,3'-Dichlorobiphenyl (PCB No. 11) 35 µg/ml in isooctane
- Formula:C₁₂H₈Cl₂
- Description:3,3'-Dichlorobiphenyl (PCB No. 11)