Organic reference standard, 2-Chlorobiphenyl (PCB No. 1) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-1-CS
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:2051-60-7
- Description:2-Chlorobiphenyl (PCB No. 1) 100 µg/ml in isooctane
- Formula:C₁₂H₉Cl
- Description:2-Chlorobiphenyl (PCB No. 1)