Organic reference standard, 3,4',5-Trichlorobiphenyl (PCB No. 39) 35 µg/ml in isooctane, CIL
Katalogové číslo PROOCIL-PCB-39
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:38444-88-1
- Description:3,4',5-Trichlorobiphenyl (PCB No. 39) 35 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:3,4',5-Trichlorobiphenyl (PCB No. 39)