Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, 3,4',5-Trichlorobiphenyl (PCB No. 39) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-39-CS
 :  
Organic reference standard, 3,4',5-Trichlorobiphenyl (PCB No. 39) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-39-CS
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    38444-88-1
  • Description:
    3,4',5-Trichlorobiphenyl (PCB No. 39) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₇Cl₃
  • Description:
    3,4',5-Trichlorobiphenyl (PCB No. 39)