Organic reference standard, 3,4,4',5-Tetrachlorobiphenyl (PCB No. 81) 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20008100IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:70362-50-4
- Description:3,4,4',5-Tetrachlorobiphenyl (PCB No. 81) 10 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:3,4,4',5-Tetrachlorobiphenyl (PCB No. 81)