Organic reference standard, 2,4',6-Trichlorobiphenyl (PCB No. 32) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-32
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:38444-77-8
- Description:2,4',6-Trichlorobiphenyl (PCB No. 32) 35 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:2,4',6-Trichlorobiphenyl (PCB No. 32)