Organic reference standard, 2,4',5-Trichlorobiphenyl (PCB No. 31) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20003100IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:16606-02-3
- Description:2,4',5-Trichlorobiphenyl (PCB No. 31) 10 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:2,4',5-Trichlorobiphenyl (PCB No. 31)