Organic reference standard, 2,4,4'-Trichlorobiphenyl (PCB No. 28) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-28-CS
Dodavatel: LGC Standards PROMOCHEM
Číslo CAS:
Specifikace
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:7012-37-5
- Description:2,4,4'-Trichlorobiphenyl (PCB No. 28) 100 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:2,4,4'-Trichlorobiphenyl (PCB No. 28)