Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, 2,4,4',5-Tetrachlorobiphenyl (PCB No. 74) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-74-CS
CAS Number:  
Organic reference standard, 2,4,4',5-Tetrachlorobiphenyl (PCB No. 74) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-74-CS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    32690-93-0
  • Description:
    2,4,4',5-Tetrachlorobiphenyl (PCB No. 74) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₆Cl₄
  • Description:
    2,4,4',5-Tetrachlorobiphenyl (PCB No. 74)

Specifications

About this item