Order Entry
Czech Republic
ContactUsLinkComponent
 
L(+)-Glutamic acid 98.5-101.5% USP, Multi-Compendial, J.T.Baker®
Catalog # 2077-06
Supplier:  Avantor
CAS Number:  
undefined
L(+)-Glutamic acid 98.5-101.5% USP, Multi-Compendial, J.T.Baker®
Catalog # 2077-06
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Assay (C₅H₉NO₄) (dried basis):
    98.5 - 100.5 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Specific Rotation [Å]²º^D (+):
    30.5 - 32.5 Degree
  • EP/BP - Ninhydrin-Positive Substances:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤200 ppm
  • EP/BP - Sulfate (SO₄):
    ≤300 ppm
  • EP/BP - Ammonium (NH₄):
    ≤200 ppm
  • EP/BP - Iron (Fe):
    ≤10 ppm
  • EP/BP - Loss on Drying:
    ≤0.5 %
  • EP/BP - Ash (sulfated):
    ≤0.1 %
  • USP - Assay (C₅H₉NO₄) (dried basis):
    98.5 - 101.5 %
  • USP - Residue on Ignition:
    ≤0.1 %
  • USP - Chloride (Cl):
    ≤0.02 %
  • USP - Sulfate (SO₄):
    ≤0.02 %
  • USP - Iron (Fe):
    ≤10 µg/g
  • USP - Related Compounds - Individual Impurities:
    ≤0.5 %
  • USP - Related Compounds - Total Impurities:
    ≤2.0 %
  • USP - Specific Rotation [Å]²º^D (+):
    31.5 - 32.5 Degree
  • USP - Loss on Drying at 105°C:
    ≤0.1 %
  • USP - Identification:
    Passes Test
  • Endotoxin Concentration, EU/g:
    ≤3.0
  • No Class 1,2,3 or other solvents are used or produced in the:
    ≤ 0.1 %
  • manufacturing or purification of the product.:
    Passes Test

Specification Test Results

About this item

 :
 :147.13 g/mol
Merck Index:13,04482
Boiling Pt:325 °C (1013 hPa)
Melting Pt:160 °C
Density:1.538 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-293-7
CAS Number:56-86-0
Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
MDL Number:MFCD00002634