Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, 2,3,4,4'-Tetrachlorobiphenyl (PCB No. 60) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-60-CS
 :  
Organic reference standard, 2,3,4,4'-Tetrachlorobiphenyl (PCB No. 60) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-60-CS
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    33025-41-1
  • Description:
    2,3,4,4'-Tetrachlorobiphenyl (PCB No. 60) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₆Cl₄
  • Description:
    2,3,4,4'-Tetrachlorobiphenyl (PCB No. 60)