Organic reference standard, 2,3,4,4',5-Pentachlorobiphenyl (PCB No. 114) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-114
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:74472-37-0
- Description:2,3,4,4',5-Pentachlorobiphenyl (PCB No. 114) 35 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,3,4,4',5-Pentachlorobiphenyl (PCB No. 114)