Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, 3,3',4-Trichlorobiphenyl (PCB No. 35) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-35
 :  
Organic reference standard, 3,3',4-Trichlorobiphenyl (PCB No. 35) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-35
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    37680-69-6
  • Description:
    3,3',4-Trichlorobiphenyl (PCB No. 35) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₇Cl₃
  • Description:
    3,3',4-Trichlorobiphenyl (PCB No. 35)