Organic reference standard, 3,3',4,5-Tetrachlorobiphenyl (PCB No. 78) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-78
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:70362-49-1
- Description:3,3',4,5-Tetrachlorobiphenyl (PCB No. 78) 35 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:3,3',4,5-Tetrachlorobiphenyl (PCB No. 78)