Organic reference standard, 2,3,3',4,5-Pentachlorobiphenyl (PCB No. 106) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-106
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:70424-69-0
- Description:2,3,3',4,5-Pentachlorobiphenyl (PCB No. 106) 35 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,3,3',4,5-Pentachlorobiphenyl (PCB No. 106)