Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, 2,3,3',5,5'-Pentachlorobiphenyl (PCB No. 111) 100 µg/ml in isooctane, CIL
Organic reference standard, 2,3,3',5,5'-Pentachlorobiphenyl (PCB No. 111) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-111-CS
CAS Number:  
Organic reference standard, 2,3,3',5,5'-Pentachlorobiphenyl (PCB No. 111) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-111-CS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    39635-32-0
  • Description:
    2,3,3',5,5'-Pentachlorobiphenyl (PCB No. 111) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,3,3',5,5'-Pentachlorobiphenyl (PCB No. 111)

Specifications

About this item